missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kallikrein 3/PSA Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF1344
This item is not returnable.
View return policy
Description
Kallikrein 3/PSA Polyclonal specifically detects Kallikrein 3/PSA in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| Kallikrein 3/PSA | |
| Polyclonal | |
| Biotin | |
| P07288 | |
| KLK3 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 3 (R&D Systems, Catalog # 1344-SE) Ala18-Pro261 Accession # P07288 | |
| 50 μg | |
| Primary | |
| Detects human KLK3 in ELISAs and Western blots. In sandwich immunoassays, less than 0.5% cross-reactivity with recombinant human (rh) KLK-1 and rhKLK-2 is observed. | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| APS seminin, EC 3.4.21, EC 3.4.21.77, Gamma-seminoprotein, hK3, kallikrein 3, (prostate specific antigen), Kallikrein-3, kallikrein-related peptidase 3, KLK2A1, KLK3, P-30 antigen, prostate specific antigen, prostate-specific antigen, PSA, PSA semenogelase, Semenogelase, Seminin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 354 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction