missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kallikrein 3/PSA Antibody, R&D Systems™

Mouse Monoclonal Antibody
£180.00 - £1530.00
Specifications
| Antigen | Kallikrein 3/PSA |
|---|---|
| Clone | 181823 |
| Concentration | LYOPH |
| Dilution | Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18734519
![]() |
R&D Systems
MAB1344-SP |
25 μg |
£180.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18718699
![]() |
R&D Systems
MAB1344-100 |
100 μg |
£387.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18760988
![]() |
R&D Systems
MAB1344-500 |
500 μg |
£1530.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kallikrein 3/PSA Monoclonal specifically detects Kallikrein 3/PSA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.Specifications
| Kallikrein 3/PSA | |
| LYOPH | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| P07288 | |
| 354 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 3/PSA Ala18-Pro261 Accession # P07288 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Kallikrein 3/PSA in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Kallikrein 5 or 11 is observed. |
| 181823 | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APS seminin, EC 3.4.21, EC 3.4.21.77, Gamma-seminoprotein, hK3, kallikrein 3, (prostate specific antigen), Kallikrein-3, kallikrein-related peptidase 3, KLK2A1, KLK3, P-30 antigen, prostate specific antigen, prostate-specific antigen, PSA, PSA semenogelase, Semenogelase, Seminin | |
| KLK3 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title