missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kallikrein 3/PSA Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB13442-500
This item is not returnable.
View return policy
Description
Kallikrein 3/PSA Monoclonal specifically detects Kallikrein 3/PSA in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| Kallikrein 3/PSA | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P07288 | |
| KLK3 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 3/PSA Pro19-Pro261 Accession # P07288 | |
| 500 μg | |
| Primary | |
| Detects human Kallikrein 3/PSA in sandwich immunoassays and in Western blots. | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 181811 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APS seminin, EC 3.4.21, EC 3.4.21.77, Gamma-seminoprotein, hK3, kallikrein 3, (prostate specific antigen), Kallikrein-3, kallikrein-related peptidase 3, KLK2A1, KLK3, P-30 antigen, prostate specific antigen, prostate-specific antigen, PSA, PSA semenogelase, Semenogelase, Seminin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 354 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction