missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kallikrein 3/PSA Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1344-SP
This item is not returnable.
View return policy
Description
Kallikrein 3/PSA Polyclonal specifically detects Kallikrein 3/PSA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.
Specifications
| Kallikrein 3/PSA | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APS seminin, EC 3.4.21, EC 3.4.21.77, Gamma-seminoprotein, hK3, kallikrein 3, (prostate specific antigen), Kallikrein-3, kallikrein-related peptidase 3, KLK2A1, KLK3, P-30 antigen, prostate specific antigen, prostate-specific antigen, PSA, PSA semenogelase, Semenogelase, Seminin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 354 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.5-1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
| P07288 | |
| KLK3 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 3 Ala18-Pro261 Accession # P07288 | |
| 25 μg | |
| Primary | |
| Detects human KLK3 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) KLK2 is observed and less than 5% cross-reactivity with rhKLK1, rhKLK4, rhKLK5, rhKLK7, rhKLK8, rhKLK9, rhKLK10, rhKLK11, rhKLK12, rhKLK13, and rhKLK15 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction