missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kallikrein 14 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2626
This item is not returnable.
View return policy
Description
Kallikrein 14 Polyclonal specifically detects Kallikrein 14 in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| Kallikrein 14 | |
| Polyclonal | |
| Unconjugated | |
| AAD50773 | |
| KLK14 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 14 Gln19-Met248, predicted Accession # AAD50773 | |
| 100 μg | |
| Primary | |
| Detects human Kallikrein 14 in ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation, Neutralization, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| EC 3.4.21, EC 3.4.21.-, hK14, kallikrein 14, Kallikrein-like protein 6, kallikrein-related peptidase 14, KLK14, KLKL6, KLK-L6kallikrein-14 | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| 43847 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction