missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kallikrein 13 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2625-SP
This item is not returnable.
View return policy
Description
Kallikrein 13 Monoclonal specifically detects Kallikrein 13 in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.
Specifications
| Kallikrein 13 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp586J1923, EC 3.4.21, EC 3.4.21.-, kallikrein 13, Kallikrein-like protein 4, kallikrein-related peptidase 13, KLK13, KLK-L4kallikrein-13, KLKL4kallikrein-like gene 4 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 26085 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunoprecipitation, Neutralization | |
| 341419 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 2.4 ug/mL | |
| Q9UKR3 | |
| KLK13 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 13 Val19-Ile262 Accession # Q9UKR3 | |
| 25 μg | |
| Primary | |
| Detects human Kallikrein 13 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Kallikrein 3, 4, 5, 6, 7, 8, 9, 10, 11, 12, 14, 15, rhKallikrein B1, rhFactorVII, rhHGFA, rhUPA, rhThrombin, rhFactor X, or rhFactor XI is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction