missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kallikrein 10 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF2007
Additional Details : Weight : 0.00970kg
Description
Kallikrein 10 Polyclonal specifically detects Kallikrein 10 in Human samples. It is validated for Western Blot.Specifications
Kallikrein 10 | |
Polyclonal | |
Unconjugated | |
O43240 | |
KLK10 | |
Mouse myeloma cell line NS0-derived recombinant human Kallikrein 10 Ala34-Asn276 Accession # O43240 | |
100 μg | |
Primary | |
Detects human Kallikrein 10 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) rhKallikrein 3, rhKallikrein 5, and rhKallikrein 11 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
breast normal epithelial cell associated serine protease, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.74, Kallikrein 10, kallikrein-related peptidase 10, KLK10, NES1, NES1kallikrein-10, Normal epithelial cell-specific 1, Protease serine-like 1, PRSSL1protease, serine-like, 1 | |
Goat | |
Affinity Purified | |
RUO | |
5655 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only