missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Kallikrein 1 (aa 130-154) Control Fragment Recombinant Protein

Product Code. 30201400
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201400

Brand: Invitrogen™ RP110083

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (68%), Rat (68%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. This protein is functionally conserved in its capacity to release the vasoactive peptide, Lys-bradykinin, from low molecular weight kininogen.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P06870
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3816
Name Human Kallikrein 1 (aa 130-154) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 0610007D04Rik; 7 S nerve growth factor gamma chain; 7 S nerve growth factor gamma subunit; esterase B; Gamma-NGF; glandular kallikrein 1; glandular kallikrein K1; Glandular kallikrein K3; glandular kallikrein-7, submandibular/renal; hK1; Kal; KALA; KAL-B; kallikrein 1; Kallikrein 1 renal/pancreas/salivary; kallikrein 1, renal/pancreas/salivary; kallikrein 1-like peptidase; kallikrein 1-related peptidase b3; kallikrein 1-related peptidase C7; kallikrein 5; kallikrein 6; kallikrein 7; kallikrein renal/pancreas/salivary; kallikrein serine protease 1; kallikrein-1; Kallikrein-related protein K1; kidney/pancreas/salivary gland kallikrein; Klk1; Klk-1; Klk1b3; Klk1b6; Klk1c1; Klk1c7; Klk1l; Klk3; Klk-3; Klk5; Klk6; Klk-6; Klk7; Klk-7; KLKR; mGk-3; mGK-6; Nerve growth factor gamma chain; Nerve growth factor gamma chain 1; Nerve growth factor gamma chain 2; nerve growth factor gamma polypeptide; nerve growth factor, gamma; Ngfg; Pancreatic kallikrein; proteinase A; PS kallikrein; RATKALA; Renal kallikrein; RGK-1; RGK-7; rK-1; RSKG-7; Tissue kallikrein; tissue kallikrein-3; Tissue kallikrein-6; True kallikrein; True tissue kallikrein
Common Name Kallikrein 1
Gene Symbol KLK1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ADTITDAVKVVELPTEEPEVGSTCL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.