missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human KAL1 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP1263
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Anosmin Synthetic Peptide, PEP-1263, from Invitrogen.
This peptide corresponds to 18 amino acids near the center of human Anosmin.PEP-1263 can be used as a blocking peptide with polyclonal antibody PA5-21149.Specifications
3730 | |
purified | |
Human KAL1 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
P23352 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
ANOS1 | |
0.02% sodium azide | |
50 μg | |
PBS |