missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human JWA (aa 129-188) Control Fragment Recombinant Protein

Product Code. 30208563
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30208563 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30208563 Supplier Invitrogen™ Supplier No. RP91079

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53319 (PA5-53319. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

JWA protein expression is affected by vitamin A and may be associated with the cytoskeleton. A protein similar to JWA in rats may play a role in the regulation of cell differentiation by inhibiting the cell membrane glutamate transporter EAAC1. The expression of the rat gene is upregulated by retinoic acid, which results in a specific reduction in EAAC1-mediated glutamate transport.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75915
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10550
Name Human JWA (aa 129-188) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5930404D22Rik; addicsin; addiscin; ADP ribosylation factor like GTPase 6 interacting protein 5; ADP-ribosylation factor GTPase 6 interacting protein 5; ADP-ribosylation factor like GTPase 6 interacting protein 5; ADP-ribosylation factor-like 6 interacting protein 5; ADP-ribosylation factor-like protein 6-interacting protein 5; ADP-ribosylation-like factor 6 interacting protein 5; Aip5; Aip-5; ARL-6-interacting protein 5; Arl6ip5; AV001879; cytoskeleton related vitamin A responsive protein; cytoskeleton-related vitamin A-responsive protein; dermal papilla derived protein 11; dermal papilla-derived protein 11; DERP11; glutamate transporter EAAC1 interacting protein; glutamate transporter EAAC1-interacting protein; glutamate transporter EEAC1-associated protein; GTRAP3-18; hp22; HSPC127; JM5; jmx; Jwa; PRA1 domain family 3; PRA1 family protein 3; Pra2; Praf3; Prenylated Rab acceptor protein 2; protein JWa; putative MAPK activating protein PM27; putative MAPK-activating protein PM27; Yip6b
Common Name JWA
Gene Symbol Arl6ip5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PLLLMFIHASLRLRNLKNKLENKMEGIGLKRTPMGIVLDALEQQEEGINRLTDYISKVKE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.