missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human JRKL (aa 414-483) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61174 (PA5-61174. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The function of this gene has not yet been defined, however, the encoded protein shares similarity with the human (41% identical) and mouse (34% identical) jerky gene products. This protein may act as a nuclear regulatory protein. Alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, May 2012].
Specifications
Specifications
| Accession Number | Q9Y4A0 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 8690 |
| Name | Human JRKL (aa 414-483) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | HHMJG; Human homolog of mouse jerky gene protein; jerky protein homolog-like; JRKL; JRK-like |
| Common Name | JRKL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction