missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human JARID1C (aa 248-333) Control Fragment Recombinant Protein

Product Code. 30210777
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210777

Brand: Invitrogen™ RP106111

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83807 (PA5-83807. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Serine/threonine kinase which acts as a negative regulator of Ras-related Rap2-mediated signal transduction to control neuronal structure and AMPA receptor trafficking. Required for normal synaptic density, dendrite complexity, as well as surface AMPA receptor expression in hippocampal neurons. Can activate the JNK and MAPK14/p38 pathways and mediates stimulation of the stress-activated protein kinase MAPK14/p38 MAPK downstream of the Raf/ERK pathway. Phosphorylates: TANC1 upon stimulation by RAP2A, MBP and SMAD1. Has an essential function in negative selection of thymocytes, perhaps by coupling NCK1 to activation of JNK1.; Isoform 4 can activate the JNK pathway. Involved in the regulation of actin cytoskeleton reorganization, cell-matrix adhesion, cell-cell adhesion and cell migration.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P41229
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8242
Name Human JARID1C (aa 248-333) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias D930009K15Rik; DXS1272E; Histone demethylase JARID1C; JARID1C; JmjC domain-containing protein SMCX; jumonji, AT rich interactive domain 1 C (Rbp2 like); Jumonji, AT rich interactive domain 1 C (RBP2-like); Jumonji/ARID domain-containing protein 1 C; KDM5C; Kiaa0234; lysine (K)-specific demethylase 5 C; lysine demethylase 5 C; lysine-specific demethylase 5 C; mKIAA0234; MR x 13; MRXJ; MRXSCJ; MRXSJ; Protein SmcX; Protein Xe169; RGD1560601; sele; selected mouse cDNA on the X; SMCX; Smcx homolog, x chromosome; Smcy homolog, X-linked; XE169
Common Name JARID1C
Gene Symbol KDM5C
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LGLMAKDKTLRKKDKEGPECPPTVVVKEELGGDVKVESTSPKTFLESKEELSHSPEPCTKMTMRLRRNHSNAQFIESYVCRMCSRG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.