missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human JAM-B/VE-JAM Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB10742T-100UG
This item is not returnable.
View return policy
Description
JAM-B/VE-JAM Monoclonal specifically detects JAM-B/VE-JAM in Human samples. It is validated for Flow Cytometry.
Specifications
| JAM-B/VE-JAM | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide. | |
| C21orf43, CD322, CD322 antigen, JAM2, JAMB, JAM-BJAM-IT/VE-JAM, junctional adhesion molecule 2JAM-2, junctional adhesion molecule B, PRO245, Vascular endothelial junction-associated molecule, VE-JAM, VE-JAMVEJAMCD322 | |
| Mouse myeloma cell line NS0-derived recombinant human JAM-B/VE-JAM Phe29-Asn236 Accession # P57087 | |
| 100 μg | |
| 58494 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 988901 | |
| Flow Cytometry 0.25-1 μg/10^6 cells | |
| P57087 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human JAM-B/VE-JAM in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction