missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human JAM-A Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1103-SP
This item is not returnable.
View return policy
Description
JAM-A Monoclonal specifically detects JAM-A in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.
Specifications
| JAM-A | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Q9Y624 | |
| F11R | |
| Mouse myeloma cell line NS0-derived recombinant human JAM-A Ser28-Ala242 (predicted) Accession # Q9Y624 | |
| 25 μg | |
| Primary | |
| Detects human JAM-A in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) JAM-B, recombinant mouse (rm) JAM-A, or rmJAM-4 is observed. | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
| 654806 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD321, CD321 antigen, F11 receptor, F11R, JAM-1, JAM1CD321, JAMA, JAM-A, JCAMJAM, Junctional adhesion molecule 1Platelet F11 receptor, junctional adhesion molecule A, PAM-1, PAM-1KAT, Platelet adhesion molecule 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 50848 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction