missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IVNS1ABP (aa 343-472) Control Fragment Recombinant Protein

Product Code. 30209500
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30209500

Brand: Invitrogen™ RP102250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51926 (PA5-51926. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

IVNS1ABP is a protein coding gene involved in many cell functions, including pre-mRNA splicing, the aryl hydrocarbon receptor (AHR) pathway, F-actin organization and protein ubiquitination. Plays a role in the dynamic organization of the actin skeleton as a stabilizer of actin filaments by association with F-actin through Kelch repeats. Protects cells from cell death induced by actin destabilization. Functions as modifier of the AHR/Aryl hydrocarbon receptor pathway increasing the concentration of AHR available to activate transcription. In addition, functions as a negative regulator of BCR(KLHL20) E3 ubiquitin ligase complex to prevent ubiquitin-mediated proteolysis of PML and DAPK1, two tumor suppressors. Inhibits pre-mRNA splicing (in vitro).; (Microbial infection) involved in the alternative splicing of influenza A virus M1 mRNA through interaction with HNRNPK, thereby facilitating the generation of viral M2 protein.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y6Y0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10625
Name Human IVNS1ABP (aa 343-472) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1190004M08Rik; 1700126I16Rik; AA960440; ARA3; aryl hydrocarbon receptor-associated 3; aryl hydrocarbon receptor-associated protein 3; FLARA3; HSPC068; influenza virus NS1A binding protein; influenza virus NS1A-binding protein; Influenza virus NS1A-binding protein homolog; Ivns1abp; kelch family protein Nd1; kelch family protein Nd1-L; kelch-like family member 39; Kelch-like protein 39; KIAA0850; KLHL39; mKIAA0850; NCX downstream gene 1; ND1; Nd1L; Nd1-L; ND1-L2; Nd1S; Nd1-S; NS1; NS-1; NS1-binding protein; NS1-binding protein homolog; Ns1bp; NS1-BP
Common Name IVNS1ABP
Gene Symbol IVNS1ABP
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QDELIEKPMSPMQYARSGLGTAEMNGKLIAAGGYNREECLRTVECYNPHTDHWSFLAPMRTPRARFQMAVLMGQLYVVGGSNGHSDDLSCGEMYDSNIDDWIPVPELRTNRCNAGVCALNGKLYIVGGSD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.