missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ITSN1 (aa 940-1039) Control Fragment Recombinant Protein

Product Code. 30196226
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30196226

Brand: Invitrogen™ RP91801

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53702 (PA5-53702. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a cytoplasmic membrane-associated protein that indirectly coordinates endocytic membrane traffic with the actin assembly machinery. In addition, the encoded protein may regulate the formation of clathrin-coated vesicles and could be involved in synaptic vesicle recycling. This protein has been shown to interact with dynamin, CDC42, SNAP23, SNAP25, SPIN90, EPS15, EPN1, EPN2, and STN2. Multiple transcript variants encoding different isoforms have been found for this gene, but the full-length nature of only two of them have been characterized so far.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15811
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6453
Name Human ITSN1 (aa 940-1039) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA517634; AA545208; AI316805; AI839402; AI848451; EH and SH3 domains protein 1; EH domain and SH3 domain regulator of endocytosis 1; Eh domain, SH3 domain regulator of endocytosis 1; Ehsh1; Ese1; human intersectin-SH3 domain-containing protein SH3P17; intersectin (SH3 domain protein 1 A); intersectin 1; intersectin 1 (SH3 domain protein 1 A); intersectin 1 (SH3 domain protein); intersectin 1 short form variant 3; intersectin 1 short form variant, 11; intersectin 1; intersectin-1; intersectin short form 2 variant 2; intersectin short variant 12; intersectin-1; intersectin-L; Itsn; ITSN1; MGC134948; MGC134949; SH3 domain-containing protein 1 A; SH3D1A; SH3P17; Src homology 3 domain-containing protein
Common Name ITSN1
Gene Symbol ITSN1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ITVLEQQDMWWFGEVQGQKGWFPKSYVKLISGPIRKSTSMDSGSSESPASLKRVASPAAKPVVSGEEFIAMYTYESSEQGDLTFQQGDVILVTKKDGDWW
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis