missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ITIH2 (aa 172-267) Control Fragment Recombinant Protein

Product Code. 30197152
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30197152

Brand: Invitrogen™ RP101364

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The inter-alpha-trypsin inhibitors (ITI) are a family of structurally related plasma serine protease inhibitors involved in extracellular matrix stabilization and in prevention of tumor metastasis. The ITI family contains multiple proteins made up of a light chain (see MIM 176870) and a variable number of heavy chains (Salier et al., 1987 [PubMed 2446322]; Himmelfarb et al., 2004 [PubMed 14744536]). ITI-HC2 may act as a carrier of hyaluronan in serum or as a binding protein between hyaluronan and other matrix protein, including those on cell surfaces in tissues to regulate the localization, synthesis and degradation of hyaluronan which are essential to cells undergoing biological processes.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P19823
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3698
Name Human ITIH2 (aa 172-267) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI747202; H2P; HC2; IGHEP2; inter-alpha (globulin) inhibitor H2; inter-alpha (globulin) inhibitor, H2 polypeptide; inter-alpha trypsin inhibitor, heavy chain 2; inter-alpha-inhibitor heavy chain 2; inter-alpha-trypsin inhibitor complex component II; inter-alpha-trypsin inhibitor heavy chain 2; inter-alpha-trypsin inhibitor heavy chain H2; Intin2; ITI heavy chain H2; ITIH2; Itih-2; ITI-HC2; PSMA1; Serum-derived hyaluronan-associated protein; SHAP
Common Name ITIH2
Gene Symbol ITIH2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LPGAKVQFELHYQEVKWRKLGSYEHRIYLQPGRLAKHLEVDVWVIEPQGLRFLHVPDTFEGHFDGVPVISKGQQKAHVSFKPTVAQQRICPNCRET
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.