missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ITIH1 (aa 605-682) Control Fragment Recombinant Protein

Product Code. 30180795
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30180795

Brand: Invitrogen™ RP98330

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (73%), Rat (73%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The inter-alpha-trypsin inhibitors (ITI) are a family of structurally related plasma serine protease inhibitors involved in extracellular matrix stabilization and in prevention of tumor metastasis. The ITI family contains multiple proteins made up of a light chain (see MIM 176870) and a variable number of heavy chains (Salier et al., 1987 [PubMed 2446322]; Himmelfarb et al., 2004 [PubMed 14744536]).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P19827
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3697
Name Human ITIH1 (aa 605-682) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias CYCSP39; H1P; HCP39; IATIH; IGHEP1; inter-alpha (globulin) inhibitor, H1 polypeptide; inter-alpha trypsin inhibitor, heavy chain 1; inter-alpha-inhibitor heavy chain 1; inter-alpha-trypsin inhibitor complex component III; inter-alpha-trypsin inhibitor heavy chain 1; inter-alpha-trypsin inhibitor heavy chain H1; Intin1; ITI heavy chain H1; ITIH; Itih1; Itih-1; ITI-HC1; MGC126415; Serum-derived hyaluronan-associated protein; SHAP
Common Name ITIH1
Gene Symbol ITIH1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PLTSMSIRGMADQDGLKPTIDKPSEDSPPLEMLGPRRTFVLSALQPSPTHSSSNTQRLPDRVTGVDTDPHFIIHVPQK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.