missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ISLR-2 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF4526
This item is not returnable.
View return policy
Description
ISLR-2 Polyclonal specifically detects ISLR-2 in Human samples. It is validated for Western Blot.
Specifications
| ISLR-2 | |
| Polyclonal | |
| Unconjugated | |
| Q6UXK2 | |
| ISLR2 | |
| Mouse myeloma cell line NS0-derived recombinant human ISLR-2 Cys20-Leu590 Accession # Q6UXK2 | |
| 100 μg | |
| Primary | |
| Detects human ISLR-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human ISLR-2 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| immunoglobulin superfamily containing leucine-rich repeat 2, ISLR2, KIAA1465, leucine-rich repeat domain and immunoglobulin domain-containing axon extension protein, LINX, UNQ1885/PRO4329 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 57611 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction