missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ISG15/UCRP APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC8044A
This item is not returnable.
View return policy
Description
ISG15/UCRP Monoclonal specifically detects ISG15/UCRP in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| ISG15/UCRP | |
| Monoclonal | |
| APC | |
| P05161 | |
| ISG15 | |
| Full-length recombinant human ISG15/UCRP Accession # P05161 | |
| 100 Tests | |
| 9636 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 851701 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| G1P2, G1P2interferon-induced 17-kDa/15-kDa protein, IFI15, IFI15hUCRP, interferon, alpha-inducible protein (clone IFI-15K), Interferon-induced 15 kDa protein, Interferon-induced 17 kDa protein, interferon-stimulated protein, 15 kDa, IP17, ISG15 ubiquitin-like modifier, Ubiquitin cross-reactive protein, ubiquitin-like protein ISG15, UCRP, UCRPIP17 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human ISG15/UCRP in Elisa and Western Blot. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction