missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ISG15/UCRP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4845
This item is not returnable.
View return policy
Description
ISG15/UCRP Monoclonal specifically detects ISG15/UCRP in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ISG15/UCRP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| ABW03539 | |
| ISG15 | |
| E. coli-derived recombinant human ISG15/UCRP Gly2-Gly157 Accession # ABW03539 | |
| 100 μg | |
| Primary | |
| Detects human ISG15/UCRP in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 539442 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| G1P2, G1P2interferon-induced 17-kDa/15-kDa protein, IFI15, IFI15hUCRP, interferon, alpha-inducible protein (clone IFI-15K), Interferon-induced 15 kDa protein, Interferon-induced 17 kDa protein, interferon-stimulated protein, 15 kDa, IP17, ISG15 ubiquitin-like modifier, Ubiquitin cross-reactive protein, ubiquitin-like protein ISG15, UCRP, UCRPIP17 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9636 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction