missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IRS2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6347
This item is not returnable.
View return policy
Description
IRS2 Monoclonal specifically detects IRS2 in Human samples. It is validated for Western Blot.
Specifications
| IRS2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS and NaN3 with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| insulin receptor substrate 2, IRS-2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8660 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 676415 | |
| Western Blot 1 ug/mL | |
| Q9Y4H2 | |
| IRS2 | |
| E. coli-derived recombinant human IRS2 Pro1035-Ile1136 Accession # Q9Y4H2 | |
| 100 μg | |
| Primary | |
| Detects human IRS2 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human IRS1 or recombinant mouse IRS3 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction