missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IRS1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB39782
This item is not returnable.
View return policy
Description
IRS1 Monoclonal specifically detects IRS1 in Human samples. It is validated for Western Blot.
Specifications
| IRS1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P35568 | |
| IRS1 | |
| E. coli-derived recombinant human IRS1 Ala801-Ala1020 (predicted) Accession # P35568 | |
| 100 μg | |
| Primary | |
| Detects human IRS1 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) IRS1 (aa 898-1066) and no cross-reactivity with rmIRS1 (aa 13-113), rmIRS3, or recombinant human IRS2 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 614115 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HIRS-1, insulin receptor substrate 1, IRS-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3667 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction