missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IRS1 (aa 602-612) Synthetic Peptide

Product Code. 11504850
Change view
Click to view available options
Quantity:
35 μg
Unit Size:
35µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11504850 35 μg 35µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11504850 Supplier Invitrogen™ Supplier No. PEP192

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

Human IRS-1 (residues 602-612) Synthetic Peptide, PEP-192, from Invitrogen.

PEP-192 is a 14 amino acid synthetic peptide corresponding to amino acid residues 602-615 from human IRS-1. The sequence of this peptide is (amino to carboxy terminus): L H T D D G Y M P M S P G V (NH2).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, catalog # PA1-1058. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 3667
Species Human
Purification Method purified
Accession Number P35568
Common Name Human IRS1 (aa 602-612)
Concentration 0.35 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol IRS1
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 35 μg
Regulatory Status RUO - research use only
Sequence L H T D D G Y M P M S P G V(NH2)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.