missing translation for 'onlineSavingsMsg'
Learn More

Human Irisin/FNDC5 Antibody, Novus Biologicals™
GREENER_CHOICE

Mouse Monoclonal Antibody

£146.00 - £516.00

Specifications

Antigen Irisin/FNDC5
Clone 1056111
Dilution Immunocytochemistry 8-25 ug/mL
Applications Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18725985
GREENER_CHOICE
View Documents
R&D Systems
MAB94201-100
100 μg
£516.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18745885
GREENER_CHOICE
View Documents
R&D Systems
MAB94201-SP
25 μg
£146.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Irisin/FNDC5 Monoclonal antibody specifically detects Irisin/FNDC5 in nan samples. It is validated for Immunocytochemistry
Specifications

Specifications

Irisin/FNDC5
Immunocytochemistry 8-25 ug/mL
Monoclonal
Purified
RUO
Q8NAU1
252995
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
1056111
Immunocytochemistry
Unconjugated
Mouse
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
fibronectin type III domain containing 5, fibronectin type III domain-containing protein 5, FNDC5, FRCP2, FRCP2Fibronectin type III repeat-containing protein 2
Synthetic peptide covering the C-term of the Irisin peptide sequences of the FNDC5 gene.,, Glu97-Glu143, Accession # Q8NAU1
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.