missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-1 beta Recombinant Protein

Product Code. 11878781
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11878781 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11878781 Supplier Invitrogen™ Supplier No. RIL1BI

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Mature human IL-1b was expressed in Pichia pastoris, KM71H. GenBank Accession # P01584, amino acids 117 - 269. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: APVRSLNCTL. Purity is >95% by SDS-PAGE analyses. Activity: The biological activity typically falls below 83.3 pg/mL in proliferation assays of mouse D10S cells. Use: For in vitro applications. A dose-response assay should be performed to determine the optimal concentration for specific experiments. Reconstitution and Dilution: Store lyophilized material at 4°C. Reconstitute in 0.5 mL of distilled or deionized water. Dilute in a buffer-containing carrier protein, aliquot, and store at or below -70°C, avoiding multiple freeze-thaw cycles. PBS with 1% BSA or a neutral media containing <10% FCS are suitable buffers.

Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01584
For Use With (Application) Control, ELISA, Immunoprecipitation, Western Blot
Formulation PBS with no preservative
Gene ID (Entrez) 3553
Molecular Weight (g/mol) 17.4 kDa
Name Human IL-1 beta
Quantity 20 μg
Source Yeast
Regulatory Status RUO
Gene Alias catabolin; cytokine; Hematopoietin 1 (H1); IFN beta inducing factor; il 1 b; IL 1β; IL-1; IL1 B; IL-1 beta; IL-1 precursor; IL1B; IL-1 B; IL1B1; IL1beta; IL-1 beta; IL1-BETA; IL1F2; IL1β; ILN; Interleukin; interleukin 1 beta; Interleukin 1 beta precursor; interleukin 1, beta; interleukin 1, beta 1; interleukin 1-beta; interleukin*1*beta; Interleukin1 beta; interleukin-1 beta; interleukin-1 beta precursor; interleukin-1 beta precursor (AA -113 to 153); interleukin-1 beta proprotein; interleukin-1 b; interleukin-1 beta; interleukin-beta; LAF; lymphocyte proliferation-potentiating factor; Osteoclast activating factor (OAF); preinterleukin 1 beta; Pro interleukin 1 beta; prointerleukin-1 beta; pro-interleukin-1-beta
Common Name IL-1 beta
Gene Symbol IL1B
Biological Activity ED50 < 83.3 pg/mL; determined by the dose-dependent proliferation of mouse D10S cells.
Conjugate Unconjugated
Species Yeast
Recombinant Recombinant
Sequence Human IL-1 beta, amino acids 117-269 (Accession # P01584)
Content And Storage 4°C
Expression System P. pastoris
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.