missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin beta 2/CD18 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB1730
This item is not returnable.
View return policy
Description
Integrin beta 2/CD18 Monoclonal specifically detects Integrin beta 2/CD18 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| Integrin beta 2/CD18 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| AAA59490 | |
| ITGB2 | |
| Mouse myeloma cell line NS0-derived recombinant human Integrin beta 2/CD18 Gln23-Asn700 Accession # AAA59490 | |
| 500 μg | |
| Primary | |
| Detects human Integrin beta 2/CD18 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Integrin beta 1 or recombinant mouse Integrin alpha 5 is observed. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 212701 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 959 beta subunit precursor)10, CD18, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), ITGB2, LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, LFA-1, LFA-1 beta, MAC-1 beta, MF17, MFI7, P150, p150,95 beta | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3689 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction