missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin alpha 6/CD49f Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1350-SP
This item is not returnable.
View return policy
Description
Integrin alpha 6/CD49f Polyclonal specifically detects Integrin alpha 6/CD49f in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| Integrin alpha 6/CD49f | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6, ITGA6B, Platelet gpl, VLA-6 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3655 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| P23229 | |
| ITGA6 | |
| E. coli-derived recombinant human Integrin alpha 6/CD49f Pro93-Arg148, Ile557-Arg613 Accession # P23229 | |
| 25 μg | |
| Primary | |
| Detects human Integrin alpha 6/CD49f in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) Integrin alpha 9 is observed and less than 1% cross-reactivity with rhIntegrin alpha 2, recombinant mouse (rm) Integrin alpha 3, rhIntegrin alpha 7, and rmIntegrin alpha 7 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction