missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin alpha 4/CD49d Antibody, R&D Systems™

Mouse Monoclonal Antibody
£143.00 - £388.00
Specifications
| Antigen | Integrin alpha 4/CD49d |
|---|---|
| Clone | 599904 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18754527
![]() |
R&D Systems
MAB13541-SP |
25 μg |
£143.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18702497
![]() |
R&D Systems
MAB13541-100 |
100 μg |
£388.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin alpha 4/CD49d Monoclonal specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for ELISA.Specifications
| Integrin alpha 4/CD49d | |
| ELISA | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), ITGA4, MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4, VLA-4 subunit alpha | |
| Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha 4/CD49d, Tyr34-Gln970 (Arg591Leu & Arg878Gln), Accession No. P13612 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Integrin alpha 4/CD49d in direct ELISA. In a sandwich immunoassay, this antibody detects the human Integrin alpha 4 beta 1 heterodimer when used with the suggested detection antibody, Mouse Anti-Human Integrin beta 1 (Catalog No. MAB17784). |
| 599904 | |
| ELISA | |
| Unconjugated | |
| Human | |
| P13612 | |
| 3676.0 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title