missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin alpha 4 beta 1 (Research Grade Natalizumab Biosimilar) Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Human Monoclonal Antibody
Brand: R&D Systems FAB10603G-100UG
This item is not returnable.
View return policy
Description
Integrin alpha 4 beta 1 Monoclonal antibody specifically detects Integrin alpha 4 beta 1 in Human samples. It is validated for Flow Cytometry
Specifications
| Integrin alpha 4 beta 1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3676 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| Hu114 | |
| Flow Cytometry 0.25-1 ug/106 cells | |
| CD49d, IA4 | |
| Human Integrin alpha 4 beta 1 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction