missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin alpha 2/CD49b Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 6 publications
Brand: R&D Systems MAB1233
This item is not returnable.
View return policy
Description
Integrin alpha 2/CD49b Monoclonal specifically detects Integrin alpha 2/CD49b in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Immunocytochemistry, CyTOF-ready.
Specifications
| Integrin alpha 2/CD49b | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ITGA2 | |
| Human keratinocytes | |
| 500 μg | |
| Primary | |
| Detects human Integrin alpha 2/CD49b. | |
| Human | |
| Purified |
| Flow Cytometry, Immunoprecipitation, Immunocytochemistry, CyTOF | |
| HAS3 | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunoprecipitation, Immunocytochemistry 15-30 ug/mL, CyTOF-ready | |
| alpha-2 subunit, CD49b, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), ITGA2, VLA 2, VLA-2 alpha | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3673 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction