missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Integrin alpha 1/CD49a Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB5676-SP
This item is not returnable.
View return policy
Description
Integrin alpha 1/CD49a Monoclonal specifically detects Integrin alpha 1/CD49a in Human samples. It is validated for Western Blot.
Specifications
| Integrin alpha 1/CD49a | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| P56199 | |
| ITGA1 | |
| Chinese hamster ovary cell line CHO-derived human Integrin alpha 1/CD49a Phe29-Pro1141 Accession # P56199 | |
| 25 μg | |
| Primary | |
| Detects human Integrin alpha 1/CD49a in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with rhIntegrin alpha 2, 3, 4, 5, 6, 8, 9, 10, 11, L, M, V, X, alpha 6x1/beta 4, alpha D/beta 2, alpha 7x2/beta 1, alpha E/beta 7, or alpha 2b/beta 3 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 639508 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD49 antigen-like family member A, CD49a, CD49a antigen, integrin, alpha 1, ITGA1, Laminin and collagen receptor, very late activation protein 1, VLA-1, VLA1integrin alpha-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3672 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction