missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Insulysin/IDE Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC2496G-100UG
This item is not returnable.
View return policy
Description
Insulysin/IDE Monoclonal specifically detects Insulysin/IDE in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Insulysin/IDE | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P14735 | |
| IDE | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Insulysin/IDE Met42-Leu1019 Accession # P14735 | |
| RUO | |
| 3416 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 334501 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Abeta-degrading protease, EC 3.4.24, EC 3.4.24.56, FLJ35968, IDE, INSDEGM, Insulin protease, insulinase, insulin-degrading enzyme, Insulysin | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Insulysin/IDE in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction