missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Insulin R/CD220 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1544
This item is not returnable.
View return policy
Description
Insulin R/CD220 Polyclonal specifically detects Insulin R/CD220 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications
| Insulin R/CD220 | |
| Polyclonal | |
| Unconjugated | |
| NP_001073285 | |
| INSR | |
| Mouse myeloma cell line NS0-derived recombinant human Insulin R/CD220 His28-Lys944 Accession # NP_001073285 | |
| 100 μg | |
| Primary | |
| Detects human Insulin R/CD220 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 0.3-15 ug/mL, CyTOF-ready | |
| CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, INSR, insulin receptor, InsulinR, IR | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3643 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction