missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human INSL5/INSL7 Recombinant Protein, PeproTech®
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
250µg
25µg
500µg
5µg
Description
Recombinant Human INSL5/INSL7 hybrid is a heterodimeric, 5.2 kDa protein consisting of a 27 amino acid beta-chain linked to a 21 amino acid alpha-chain by two disulfide bonds. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
The Insulin/Relaxin structural family of proteins consists of at least seven members, each containing an insulin-like heterodimeric structure of two polypeptide chains held together by two inter-chain disulfide bonds. The INSL5/INSL7 hybrid contains the beta-chain of INSL7 (Relaxin-3), linked to the alpha-chain of INSL5. The INSL5/INSL7 hybrid protein can specifically activate the G protein-coupled receptors, GPCR135 and GPCR142, but unlike Relaxin-3, does not induce significant activation of the LGR7 receptor.
Specifications
Specifications
| Accession Number | Q8WXF3, Q9Y5Q6 |
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 10022, 117579 |
| Molecular Weight (g/mol) | 5.2 kDa |
| Name | Human INSL5/INSL7 |
| Quantity | 100 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction