missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Recognizes human INS
- Insulin is a peptide hormone, produced by beta cells of pancreas and is central to regulating carbohydrates and fat metabolism in the body
- Participates in glucose utilization, protein synthesis and formation and storage of neutral lipids
- Insulin synthesizes as precursor molecule, proinsulin, which is processed prior to secretion
- A and B peptides are joined together by a disulfide bond to form insulin, while central portion of precursor molecule is cleaved and released as C-peptide
- Defects in insulin results in Type 1 diabetes mellitus
Process:
- Antibody specific for INS pre-coated onto microwells
- INS protein in samples captured by the coated antibody after incubation
- After extensive washing, another antibody of biotinylated specific for INS is added to detect captured INS protein
- For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent
- Solution containing sulfuric acid used to stop color development and color intensity
- Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm
Specifications
Specifications
| Accession Number | NP_000198 |
| Assay Range | 7.8 to 500 pmol/L |
| Assay Sensitivity | 1.3 pmol/L |
| Certifications/Compliance | ISO 13485 |
| Detection Method | Sandwich ELISA, Double Antibody |
| Immunoassay Kit Format | Coated ELISA Kit |
| Sample Type | Plasma, Serum |
| Substrate | DAB |
| Target Species | Human |
| Gene ID (Entrez) | 3630 |
| Show More |
Safety and Handling
Product Identifier
- 96TEST AuthentiKine Human M-CSF ELISA Kit
- Warning
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P234-Keep only in original container.
- P264-Wash skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?