missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Inhibin A Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8649-SP
This item is not returnable.
View return policy
Description
Inhibin alpha Monoclonal specifically detects Inhibin alpha in Human samples. It is validated for Western Blot.
Specifications
| Inhibin alpha | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| A-inhibin subunit, INHA, Inhibin A, inhibin alpha chain, inhibin, alpha | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3623 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 58608 | |
| Western Blot 1 ug/mL | |
| P08476 | |
| INHA | |
| Peptide on N-terminus of human Inhibin A protein Accession # P08476 | |
| 25 μg | |
| Primary | |
| Detects human Inhibin A in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Activin A is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction