Learn More
BD Human Induced Pluripotent Stem Cell Analysis and Sorting Kit
For sorting and analysis of induced human pluripotent stem cell (hiPSC) cultures and cells in the process of being reprogrammed
Brand: BD 562626
Additional Details : Weight : 0.14000kg
Description
- Species Reactivity: Hu
- Size: 50 tests
Flow Cytometry
Specifications
CD13 PerCP-Cy™5.5, SSEA-4 Alexa Fluor™ 647, TRA-1-60 PE, isotype control Antibodies, BD™ compBead Plus compensation particles | |
50 Tests |