missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IMMP1L (aa 31-84) Control Fragment Recombinant Protein

Product Code. 30210965
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210965

Brand: Invitrogen™ RP108428

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84373 (PA5-84373. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The mitochondrial inner membrane peptidase (IMP) complex generates mature, active proteins in the mitochondrial intermembrane space by proteolytically removing the mitochondrial targeting presequence of nuclear-encoded proteins. IMP1 and IMP2 (IMMP2L; MIM 605977) are the catalytic subunits of the IMP complex.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96LU5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 196294
Name Human IMMP1L (aa 31-84) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1500034J20Rik; 2610528O17Rik; AU042386; IMMP1; Immp1l; IMP1; IMP1 inner mitochondrial membrane peptidase-like; IMP1 inner mitochondrial membrane peptidase-like (S. cerevisiae); IMP1-LIKE; IMP1-like protein; inner mitochondrial membrane peptidase subunit 1; mitochondrial inner membrane protease subunit 1
Common Name IMMP1L
Gene Symbol IMMP1L
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GGVVMCSGPSMEPTIQNSDIVFAENLSRHFYGIQRGDIVIAKSPSDPKSNICKR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.