missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL1RAPL1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB975-SP
This item is not returnable.
View return policy
Description
IL1RAPL1 Monoclonal specifically detects IL1RAPL1 in Human samples. It is validated for Western Blot.
Specifications
| IL1RAPL1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL1R8IL1RAPL, IL-1R9, IL-1RAPL1, IL-1-RAPL-1, IL-1RAPL-1, IL1RAPL-1, interleukin 1 receptor accessory protein-like 1, interleukin 1 receptor-8, interleukin-1 receptor accessory protein-like 1, mental retardation, X-linked 10, MRX10, MRX21, MRX34, oligophrenin-4, OPHN4mental retardation, X-linked 21, Three immunoglobulin domain-containing IL-1 receptor-related 2, TIGIRR-2, TIGIRR-2mental retardation, X-linked 34, X-linked interleukin-1 receptor accessory protein-like 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11141 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 143116 | |
| Western Blot 1 ug/mL | |
| Q9NZN1 | |
| IL1RAPL1 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-1 RAPL1/IL-1 R8 Leu19-Thr357 Accession # Q9NZN1 | |
| 25 μg | |
| Primary | |
| Detects human IL1RAPL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) IL-1 RI, rhIL-1 RII, rhIL-1 RAcP, rhIL-1 R4, rhIL-1 R5, rhIL-1 Rrp2, rhIL-1 R7, or rhIL-1 RAPL2 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction