missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL18BP (aa 113-194) Control Fragment Recombinant Protein

Product Code. 30211492
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30211492

Brand: Invitrogen™ RP102904

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (57%), Rat (57%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59412 (PA5-59412. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Interleukin-18 binding protein (IL-18BP) is a constitutively secreted glycoprotein that acts as a natural antagonist for interleukin-18 (IL-18) by preventing interaction between IL-18 and its receptors. Through this binding, IL-18BP neutralizes the proinflammatory role of IL-18 in cell-mediated immune responses, resulting in reduced IFN-gamma production by T helper type I cells and inhibition of allospecific cytotoxic T lymphocyte (CTL) activity through augmented natural killer (NK) cell cytotoxicity. As an inhibitor of IL-18 and an essential component of skin homeostasis, IL-18BP has been linked to a variety of diseases, including eczema, asthma, and the autoimmune condition, Crohn's disease. The upregulation of IL-18BP by keratinocytes expressing the human papillomavirus (HPV) E7 oncoprotein has been shown to decrease IL-18-induced IFN-gamma production and IL-18-mediated T cell activation. Elevated levels of IL-18 and IL-18BP, as well as proinflammatory cytokines IFN-gamma, TNF-alpha, IL-1beta and IL-8, have been linked to chronic inflammation and observed in macrophages (including Kupffer cells) isolated from lesions and intestinal tissues of Crohn's disease patients. As a member of the immunoglobulin-like class of receptors, IL-18BP contains a single immunoglobulin (Ig) domain; although two (b and d) of the four identified human isoforms (a-d) contain incomplete domains that greatly reduce their binding affinity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95998
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10068
Name Human IL18BP (aa 113-194) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias Igifbp; Il18bp; IL-18 BP; IL18BPa; interferon gamma inducing factor binding protein; interferon gamma-inducing factor-binding protein; interleukin 18 binding protein; interleukin-18 binding protein; interleukin-18-binding protein; MC51L-53 L-54 L homolog gene product; MC54L; Tadekinig-alfa
Common Name IL18BP
Gene Symbol IL18BP
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RLWEGSTSRERGSTGTQLCKALVLEQLTPALHSTNFSCVLVDPEQVVQRHVVLAQLWAGLRATLPPTQEALPSSHSSPQQQG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.