missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL13RA1 (aa 30-152) Control Fragment Recombinant Protein

Product Code. 30201748
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201748

Brand: Invitrogen™ RP91957

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-81870 (PA5-81870. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a subunit of the interleukin 13 receptor. This subunit forms a receptor complex with IL4 receptor alpha, a subunit shared by IL13 and IL4 receptors. This subunit serves as a primary IL13-binding subunit of the IL13 receptor, and may also be a component of IL4 receptors. This protein has been shown to bind tyrosine kinase TYK2, and thus may mediate the signaling processes that lead to the activation of JAK1, STAT3 and STAT6 induced by IL13 and IL4.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P78552
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3597
Name Human IL13RA1 (aa 30-152) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI882074; Cancer/testis antigen 19; CD antigen CD213a1; CD213a1; CD213a1 antigen; CT19; IL13 receptor alpha-1 chain; IL-13 receptor subunit alpha-1; IL13R; IL-13 R subunit alpha-1; IL-13 r[a]; IL13RA; IL-13 Ra; IL13RA1; IL-13 RA1; IL-13 R-alpha-1; interleukin 13 receptor subunit alpha 1; interleukin 13 receptor, alpha 1; interleukin 13 receptor, alpha 1-like; interleukin-13 receptor subunit alpha-1; Interleukin-13-binding protein; LOC100360218; Novel cytokine receptor 4; NR4
Common Name IL13RA1
Gene Symbol IL13RA1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ETQPPVTNLSVSVENLCTVIWTWNPPEGASSNCSLWYFSHFGDKQDKKIAPETRRSIEVPLNERICLQVGSQCSTNESEKPSILVEKCISPPEGDPESAVTELQCIWHNLSYMKCSWLPGRNT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.