missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-9R Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £474.00
Specifications
| Antigen | IL-9R |
|---|---|
| Clone | 33449 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18783378
![]() |
R&D Systems
MAB2901-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18179140
![]() |
R&D Systems
MAB2901 |
500 μg |
£474.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-9R Monoclonal specifically detects IL-9R in Human samples. It is validated for Western Blot.Specifications
| IL-9R | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD129, CD129 antigen, IL-9 R, IL-9 receptor, IL9R, IL-9R, interleukin 9 receptor, interleukin-9 receptor | |
| IL9R | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 33449 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q01113 | |
| 3581 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-9 R Ser41-Pro270 Accession # Q01113 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-9 R in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) IL-4 R rhIL-5 R alpha , rhIL-5 R beta , rhIL-13 R alpha 1, rhIL-13 R alpha 2, and recombinant mouse IL-9 R is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title