missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-8 (CXCL8) (1-77), Recombinant Protein

Product Code. p-7091800
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15537496 10 μg 10µg
15547496 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15537496 Supplier Invitrogen™ Supplier No. 14808963

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Human IL-8, also called neutrophil attractant/activating protein (NAP-1), monocyte derived neutrophil activating peptide (MONAP), monocyte derived neutrophil activating factor (ANF), monocyte derived neutrophil chemotactic factor (MDNCF), and neutophil activating factor (NAF), is a ∽6-8 kDa factor produced by variety types of cells including monocytes, lymphocytes, granulocytes, fibroblasts and endothelial cells. IL-8 is an inflammatory cytokine which functions as a neutrophil chemoatractant and activating factor. It also attracts basophils and a subpopulation of lymphocytes.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_000584
For Use With (Application) Cytokine Bioassay
Formulation Sterile liquid; phosphate buffered saline, pH 7.2, 1.0% BSA. 0.22μm filtered.
Molecular Weight (g/mol) The protein is not methionylated at the N-terminal and has a predicted molecular mass of 8,386. The DTT reduced protein migrates as an 8kDa polypeptide and the non-reduced polypeptide migrates as a 9kDa polypeptide on SDS-PAGE.
Name IL-8
Purification Method SDS-PAGE
Quantity 10 μg
Source E. coli
Storage Requirements Store at less than or equal to -70°C.
Regulatory Status RUO
Structural Form E. coli expressed amino acids Ser 28-Ser 99 of human IL-8 accession # NM_000584
Endotoxin Concentration Less than 0.01ng/μg cytokine as determined by the LAL assay.
Gene Symbol Interleukin-8, IL8, 72aa
Biological Activity Measured by chemotaxis assay of human peripheral blood neutrophils. Maximal chemoattractant activity was observed between 10-100ng/mL.
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Purity or Quality Grade >98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.