Learn More
Human IL-8/CXCL8 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 9 publications
Brand:  R&D Systems AF-208-NA
Product Code. 18041912
  • £512.00 / 100µg
Description
CXCL8/IL-8 Polyclonal specifically detects CXCL8/IL-8 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Neutralization, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CXCL8/IL-8
Polyclonal
Unconjugated
P10145
IL8
E. coli-derived recombinant human IL-8/CXCL8 Ser28-Ser99 Accession # P10145
100 μg
Primary
Detects human IL-8/CXCL8 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant canine IL-8/CXCL8, recombinant feline IL-8/CXCL8 and recombinant porcine IL-8/CXCL8 is observed.
Human
IgG
Western Blot, Immunohistochemistry, Neutralization, Immunocytochemistry
LYOPH
Western Blot 2 ug/mL, Simple Western 25 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.1-0.5 ug/mL, Immunocytochemistry 5-15 ug/mL
3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8, CXCL8SCYB8, Emoctakin, GCP1, GCP-1TSG-1, IL8, interleukin 8, K60, LAI, LECT, LUCT, LYNAP, MDNCF, MDNCFb-ENAP, member 8, MONAP, MONAPGCP1, NAF, NAP1, NAP-1NAP1, NCF, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor, TCF, TSG1
Goat
Affinity Purified
RUO
3576
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.