missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-7 Recombinant Protein

Recombinant Human Interleukin 7 (IL7) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHC0076

Code : 50

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 10068782

  • £906.00 / Each
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Human IL-7 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

IL7 stimulates the proliferation of pre-B and pro-B cells. IL7 also supports the maturation of megakaryocytes.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human PBMCs
  • Proven Compatibility: Gibco proteins are bioassayed with Gibco media

Approved for shipment at Room Temperature or on Wet or Dry Ice.

Specifications

Specifications

Cell Culture
17.5 kDa
25μg
Human
Carrier-Free
25 μg
E. coli
Recombinant, Ligand
Interleukins
Gibco™
3574
Sequential Chromatography
IL-7
Store in refrigerator (2–8°C).
< 0.1 ng/μg
Approved for shipment on Wet or Dry Ice
Cytokines & Receptors
Lyophilized
Immunology, Signal Transduction, Stem Cell Research, Drug Development, Oncology
95 %
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.