missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-7 High Sensitivity Magnetic Luminex™ Performance Assay

Product Code. 18401490
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401490 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401490 Supplier R&D Systems™ Supplier No. LBHS207

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Magnetic bead-based multiplex assay for the Luminex™ MAGPIX™ platform

  • Magnetic format allows for easier wash steps
  • Undergoes similar validation testing as Quantikine™ ELISA Kits
  • Precise and Specific
  • Requires a small sample volume (<50 μL)
  • Run your assay in just 3-5 hours
  • Mass-calibrated standards for consistent results with every new lot of material
  • Enough reagents for one 96-well plate
TRUSTED_SUSTAINABILITY

Specifications

Product Type Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform
Quantity 1 Kit
Species Human
Binding Target IL-7
Bead Type Magnetic
Assay Sensitivity 0.14pg/mL
Assay Range 0.83 - 3390pg/mL (Serum, Heparin Plasma, EDTA Plasma), 0.83 - 848pg/mL (Cell Culture Supernates)
Gene ID (Entrez) 3574
Gene Symbol IL7
Sample Type Serum, Plasma (EDTA, Heparin), Cell Culture Supernatant
Sample Volume Cell Culture Supernates (100μL),Serum (50μL),EDTA Plasma (50μL),Heparin Plasma (50μL)
Kit Contents
  • Microparticle Concentrate
  • Biotin-Antibody Concentrate
Content And Storage Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.