missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-7 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF-207-NA
This item is not returnable.
View return policy
Description
IL-7 Polyclonal specifically detects IL-7 in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| IL-7 | |
| Polyclonal | |
| Unconjugated | |
| P13232 | |
| IL7 | |
| E. coli-derived recombinant human IL-7 Asp26-His177 Accession # P13232 | |
| 100 μg | |
| Primary | |
| Detects human IL-7 in direct ELISAs and Western blots. In Western blots, detects recombinant mouse IL-7 but no cross-reactivity with recombinant rat IL-17. | |
| Human | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.02-0.04 ug/mL | |
| IL7, IL-7interleukin-7, interleukin 7, Lymphopoietin-1, PBGF | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3574 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction