missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-7, Animal-Free Recombinant Protein, PeproTech®

Product Code. 17843273
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 17843273

Brand: Gibco™ AF20007100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human IL-7 is a 17.5 kDa protein containing 153 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukin 7 (IL-7) is a hematopoietic cytokine that is constitutively produced by stromal cells from the bone marrow and thymus. IL-7 signals through the IL-7R to promote the differentiation of hematopoietic stem cells to lymphoid precursor cells, which give rise to T cells, B cells, and natural killer cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13232, P16871
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 3574, 3575
Molecular Weight (g/mol) 17.5 kDa
Name Human IL-7, Animal-Free
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias A630026I06Rik; H-IL-7; hlb368; IL7; Il-7; ILN; Interleukin; interleukin 7; Interleukin7; interleukin-7; M-IL-7
Common Name IL-7
Gene Symbol Il7
Biological Activity Determined by its ability to stimulate the proliferation of mouse 2E8 cells. The expected ED50 ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^6 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence MDCDIEGKDG KQYESVLMVS IDQLLDSMKE IGSNCLNNEF NFFKRHICDA NKEGMFLFRA ARKLRQFLKM NSTGDFDLHL LKVSEGTTIL LNCTGQVKGR KPAALGEAQP TKSLEENKSL KEQKKLNDLC FLKRLLQEIK TCWNKILMGT KEH
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.