missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-6R (soluble) Recombinant Protein, PeproTech®

Product Code. 17801073
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
50µg
5µg
This item is not returnable. View return policy

Product Code. 17801073

Brand: Gibco™ 20006RC5UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

PeproTech's CHO cell-derived Recombinant Human sIL-6 Receptor alpha is a 37.9 kDa glycoprotein corresponding to 339 amino acid residues of the extracellular domain of IL-6Ralpha. As a result of glycosylation, Recombinant Human sIL-6 Receptor alpha migrates with an apparent molecular mass of approximately 57-70 kDa by SDS-PAGE gel, under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Interleukin 6 receptor (IL6R), also known as CD126 (Cluster of Differentiation 126), is a type I cytokine receptor. This gene encodes a subunit of the interleukin 6 (IL6) receptor complex. Interleukin 6 is a potent pleiotropic cytokine that regulates cell growth and differentiation and plays an important role in the immune response. The IL6 receptor is a protein complex consisting of this protein and interleukin 6 signal transducer (IL6ST/GP130/IL6-beta), a receptor subunit also shared by many other cytokines. Dysregulated production of IL6 and this receptor are implicated in the pathogenesis of many diseases, such as multiple myeloma, autoimmune diseases and prostate cancer. Alternatively spliced transcript variants encoding distinct isoforms have been reported. A pseudogene of this gene is found on chromosome 9.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08887
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 3570
Molecular Weight (g/mol) 37.9 kDa
Name Human IL-6 R (soluble)
Quantity 5 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias CD126; CD126 antigen; gp80; IL-6 Receptor; il-6 receptor alpha; IL-6 receptor alpha chain; IL-6 receptor subunit alpha; IL6Q; IL6R; IL-6 R; IL-6 R 1; IL-6 R subunit alpha; IL6R1; IL-6 R-1; Il6ra; IL-6 RA; IL-6 R-alpha; IL6RQ; ILN; Interleukin; interleukin 6 receptor; interleukin 6 receptor, alpha; interleukin-6 receptor subunit alpha; Membrane glycoprotein 80; MGC104991
Common Name IL-6 Receptor (CD126)
Gene Symbol IL6R
Biological Activity Determined by its ability to intensify the IL-6 induced growth inhibition of mouse M1 cells. The expected ED50 is ≤ 5.0 ng/ml, in the presence of 20 ng/ml of rhIL-6.
Conjugate Unconjugated
Recombinant Recombinant
Sequence LAPRRCPAQE VARGVLTSLP GDSVTLTCPG VEPEDNATVH WVLRKPAAGS HPSRWAGMGR RLLLRSVQLH DSGNYSCYRA GRPAGTVHLL VDVPPEEPQL SCFRKSPLSN VVCEWGPRST PSLTTKAVLL VRKFQNSPAE DFQEPCQYSQ ESQKFSCQLA VPEGDSSFYI VSMCVASSVG SKFSKTQTFQ GCGILQPDPP ANITVTAVAR NPRWLSVTWQ DPHSWNSSFY RLRFELRYRA ERSKTFTTWM VKDLQHHCVI HDAWSGLRHV VQLRAQEEFG QGEWSEWSPE AMGTPWTESR SPPAENEVST PMQALTTNKD DDNILFRDSA NATSLPVQD
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.